Samplers For Liquids
Sampling equipment for taking samples out of most diverse containers and depths e.g. from small jerrycans up to big basins, tanks or open water courses.
Dip samplers and plunging siphons
Samplers for low viscosity liquids. Available in different versions, lengths and materials so the sampler can be adapted to the application.
Scoops
Made from diverse materials to scoop and sample liquids and viscous materials. Telescopic solutions help to reach distant sampling points.
Vacuum samplers
Convenient solution when samples need to be collected from depth (up to 5 m) that cannot be reached with lances due to limitation in length.
Dipping bottles
The solution when samples of liquids at considerable depth needs to be taken.