Leica Compound Inverted Microscopes
Leica inverted microscopes offer you high performance to help you advance your research.
The Leica DMi1 inverted microscope supports your specific work routine in your cell culture lab. Its operation is so intuitive, its handling so comfortable that you can fully concentrate on your work. Choose the functions you need, and, if necessary, you can easily add a variety of accessories that are important for your work. With high-performance optics, ergonomic design, and 5W LED illumination, the Leica DM IL LED is ideal for cell culture, micromanipulation, documentation of immunostained specimens, and routine live cell examinations.
For special diagnostics requirements, the microscope is certified for in-vitro-diagnostics (IVD) including in-vitro-fertilization (IVF).
DMi1
Just right for your live cell lab – smart choice for quick checks. The well designed and quality control elements enable you to work in comfort.
DM IL LED
The Leica DM IL LED inverted routine microscope features a comprehensive set of contrast methods to monitor your specimen the way you need.
Mateo TL
A digital microscope for routine cell culture with an intuitive setup and use, making the process of checking cell culture growth quick and effortless for every member in your lab.