Filtration
Dissolved foodstuffs are often very difficult to filter due to the amount of relatively large particulate material present in the solution. Depending on the application and analytical technique involved, products including folded filter papers for gravity filtration or syringe filters for positive pressure filtration can be used to clarify samples.
Product selection
Filter Papers
VWR offers an extensive line of filter papers...
Syringe Filters
Positive pressure can make filtration easier!
Glass Fibre Filters
Glass fibre filters feature fast flow rates, high loading capacity, wide thermal tolerance and excellent precipitate retention...