DuPont™ Tyvek® products
Tyvek® is a unique nonwoven fabric that comes with inherent protection that is engineered right in. There are no films, and no laminates that can abrade or wear away over time – Tyvek® maintains wearer protection during use. Designed for an optimal balance of protection, durability and comfort, Tyvek® protective apparel is ideal for a wide range of jobs. From spray painting to handling lead or asbestos abatement, the ergonomic design helps keep workers safe and effective when the going gets rough. It is also devoid of fillers or additives and is silicon-free.
Tyvek® 500 Xpert
Setting a new standard of protection in the Type 5 and 6 category through greater protection and comfort. Tyvek® garments are composed of flash spun high density polyethylene, providing an ideal balance of protection, durability and comfort.
Tyvek® 500 Labo
Protecting wearers and processes in laboratories and clean environments. Tyvek® is permeable to both air and water vapour, yet repels water-based liquids and aerosols.
Tyvek® 600 Plus
Protecting wearers and processes in laboratories and clean environments. Tyvek® is permeable to both air and water vapour, yet repels water-based liquids and aerosols.
Video
Tyvek® makes the difference
DuPont™ Tyvek® offers an ideal balance of protection, durability, and comfort.
Literature
DuPont Personal Protection
Product catalogue