Microbiology Filter Funnels
Disposable Filter Funnels with membrane, sterile
![]() |
MicroFunnel filter funnels 100 – 300 ml Ideal for quality control analysis of aqueous fluids used in pharmaceutical production. Individually labeled for lot traceability. Microcheck II™ Beverage monitors filter funnels Easy-to-use disposable filter funnel to meet microbial analysis needs for beverages. MicroFunnel Plus filter funnels 300 ml Combined filter funnel and sample cup to eliminate the risk of contamination. Products with Supor® membrane are designed to allow sampling of hot water up to 90 ºC. |
Disposable Filter Funnels without membrane, sterile
![]() |
Sentino® Filter funnels 100 – 250 ml Suitable for drinking and waste water analysis, environmental and recreational water monitoring, industrial water system monitoring, beverage monitoring. |
Reusable Filter Funnels, autoclavable
![]() |
Sentino® Magnetic filter funnels Suitable for drinking and waste water analysis, environmental and recreational water monitoring, industrial water system monitoring, beverage monitoring, pharmaceutical raw materials, intermediates, and final product. |