Bioprocessing Centrifuges
Featured Solutions
Fast and efficient separation of cell cultures from bioreactors or other sources, at high g-forces, while simplifying productivity, safety and reliability: Versatile, high-throughput sample processing 6 x 1L bottles at 17,568xg or 4 x 1L at 20,584xg.
Additional Resources
The Sorvall BIOS 16 centrifuge provides outstanding capacity to maximise high-throughput bioprocessing productivity: process from 6 x 1L bottles to 8 x 2L bottles per run at speeds up to 8,500xg!
The Sorvall BIOS A centrifuge offers high performance for application flexibility: up to 10 x 1L bottles with performance up to 12,000xg.