Cleanroom Tapes
Adhesive cleanroom tapes are suitable for color marking, marking of surfaces, equipment, machinery, small parts and they are also suitable for bar code labels. They have a strong grip, but can be removed without residue. Very low generation of particles and outgassing, core is made of plastic. Available in different colours and different widths and length.
We recommend cuff stripes for an effective seal between glove and garment. Low-tack acrylic adhesive removes cleanly and leaves no adhesive residue or damage to garments. With heavy liner for easy separation, gamma irradiated.
Product selection
Tapes
Please choose from a list of products and refine your search by attributes...