Quantus Garments
Please ask which models are available in respective fabrics and sizes.
Fabrics (Overalls, Hoods, Facemasks, Boot-/ Shoe Covers, Coats)
Model Overview
Sizing
Coats
With press studs or zipper (model: Quantus Control), with or without pockets.
Overalls
Overalls with lapped seams for a well-sealed and secure seam covering. With open face hood and facemask, as combined version with integrated flat hood or with special ESD function available.
Facemasks
Press studs or with velcro strip for attaching to the hoods
Headwear
Headwear 'Bouffant style' with elastic band for size adjustment and press studs for width adjustment.
Hoods
Optimal fit through continuous width adjustment in back, with shoulder protection and with press studs to button the facemask onto the side of the hood.
Boot Covers
Optimum fit due to adjustments at top, at instep and at knee and ankle high, with robust, waterproof and autoclavable sole.
Shoe Covers
Elastic band on upper side, width adjustment, double reinforcement. With stabil and waterproof plastic sole .