Wet Processing Equipment
Features:
  • Self-supporting design of solidly welded 20 mm polypropylene panels
  • Work panels optionally of stainless steel or natural polypropylene
  • Surface extraction suction via the perforated work panel
  • Collection troughs with air ducts, solidly welded into the casing
  • Collection trough floor with 2% gradient laterally towards the rear
  • Collection trough with coarse sieve
  • Acid bath, rinsers, rinsing bathes, etc. can be optionally integrated
  • Lower area is fitted with folding doors. The lower area can be extraction suctioned similar to the perforated working panel
  • Optionally the base can stand on height-adjustable stainless steel feet
  • Basic supply connections are on the rear side of the bench
  • Operating and display elements are easily visible on the front side at the table height
  • The transparent cover ensures protection against sprayed water
Dimensions:
Model: CT 975
Bench width: 975 mm
Raster quantity (work panels/troughs): 2
Bench depth: 695 or 850 mm
Door width: 675 mm
Model: CT 1280
Bench width: 1280 mm
Raster quantity (work panels/troughs): 3
Bench depth: 695 or 850 mm
Door width: 780 mm
Model: CT 1585
Bench width: 1585 mm
Raster quantity (work panels/troughs): 4
Bench depth: 695 or 850 mm
Door width: 1085 mm
Model: CT 1890
Bench width: 1890 mm
Raster quantity (work panels/troughs): 5
Bench depth: 695 or 850 mm
Door width: 1390 mm