Cleanroom Bucket Systems
Stainless steel Cleanroom Cleaning Equipment
Available as Single- and Double-Bucket System. Accessories are completely autoclavable (can be disassembled into individual parts) and have a GMP conformant design.
The cleanroom compatibility of the cleaning equipment is ensured through long-term practical tests. Available with mop and mop frame. All versions with Ringo flat squeezer ES-CR. Can be optionally upgraded with surface cleaning unit. Additional accessories or product-specific adaptations are available on request.
Systembox EasyMop GMP®
The patented MopJump-system with displacement area allows for any easy impregnation of the mop head as well as a contact-free pick up. The EasyMop GMP® Systembox is suitable for 10 mops. Triple sealing ensures that the box is securely closed. The MicroSicuro CR/A mops are made of special yarn suitable for cleanrooms (ISO 5). Material is developed especially for the impregnation method.
The EasyMop GMP-System Trolley is also made of stainless steel, GMP-conform, with special autoclavable wheels and with up to 4 boxes.
Product selection
Single- and Double-Bucket Systems
Please choose from a list of products and refine your search attributes...