Gas Regulators
High-Purity Single-Stage Regulators, Chrome-Plated Brass or 316 Stainless Steel
These high-purity, single-stage regulators are ideally suited for primary pressure control of noncorrosive high-purity gases (chrome-plated brass) or corrosive ultra high-purity gases (316 stainless steel). They are designed for use with high-purity carrier gases or zero-span and calibration gases in applications including gas chromatography or the control of gases from cryogenic sources.
With a stainless steel diaphragm and high leak integrity, these regulators help prevent gas contamination. Single-stage regulators are recommended as primary pressure control for applications that can tolerate minor fluctuations in outlet pressure as inlet pressure changes.
Other delivery pressures and connections are available. Contact your VWR Sales Representative for information.