Precise and reproducible dosing for food laboratories
BioTool’s DosiPump stainless smooth, steel peristaltic pump for microbiology and food laboratories.
- Simple cleaning and access to all parts
- Sturdy full metal body allows direct stage bar fixation
- Easy tube load system for quick tube exchange
- Unique metal pump head ideal for tempered or hot liquids
- Low pulsation by 2-channel, 10-roller pump head
- 2 pump versions: Up to 800 ml/min & 2400 ml/min
Log in to see your contract pricing and availability.