Bollé Safety
Bollé Safety prescription lenses - benefit from the incomparable experience of a certified laboratory, as well as expert designers and technicians. Carefully selected portfolio to bring you a wide choice of plastic and metal frames, with optimum protection. The product line features various frames that can be fitted with four types of prescription lens (unifocal, bifocal, progressive or degressive) and three different lens materials (polycarbonate, CR39 or mineral glass).
Download the Bollé brochure to find out more
All Inclusive Prescription Pack (Cat. No. BOLERXPACK) - a simple way to order up to x10 prescription safety glasses. Click to download pdf.
Bollé’s All Inclusive Prescription Pack is an easy way you to access prescription eyewear for one simple cost. Each pack contains, 10 individual prescription eyewear packets, each one containing prescription form, pre-paid voucher and dispensing fees, and gives access to the standard Bollé frames - single vision or varifocal lens.
Once purchased, the individual takes one of the packets to their local optician who will complete the form and send it to be processed. Once the frames are ready, the optician will notify the wearer for collection.