Human Factor IX/PTC ELISA Kit
Supplier: Abcam
AB300307-96 AB300307-10X96 AB300307-1X384
ABCAAB300307-96EA 758 GBP
ABCAAB300307-96 ABCAAB300307-10X96 ABCAAB300307-1X384
Human Factor IX/PTC ELISA Kit
Assays ELISAs
Human Factor IX/PTC ELISA kit is a single-wash 90-min Simplestep used to quantify Human Factor IX/PTC with a sensitivity of 15.9 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader. Different formats for different needs: 10×96 plates for bulk orders and 384-well plate for higher throughput.