HexArmor® ThornArmor® 3092 Thorn and Needle-Resistant Gloves
Supplier: UVEX
6001006 6001007 6001008 6001009 6001010 6001011
UVEX6001006EA 0 GBP
UVEX6001006 UVEX6001007 UVEX6001008 UVEX6001009 UVEX6001010 UVEX6001011
HexArmor® ThornArmor® 3092 Thorn and Needle-Resistant Gloves
Gloves
ThornArmor® 3092 gloves deliver industry-leading protection against sharp hazards, setting the hand protection standard for serious gardeners, hobbyists, and professional landscapers.
  • High needlestick resistance provided by SuperFabric® brand material
  • Breathable back-of-hand
  • Durable, protective palm material
  • Launderable
SuperFabric® brand material protects against hazards through a configuration of tiny guard plates layered onto performance fabrics. This exclusive material offers pinprick protection and extremely high cut protection. These gloves feature three layers of SuperFabric® *brand material on the palm (interior layer) and a reinforced index finger wrap extending the puncture-resistant area. Durable TP-X® palm provides superior grip and abrasion resistance. A breathable HexVent® panel on the back-of-hand helps keep wearers cool, while the form-fitting design increases dexterity and comfort on the job.