Anti-Aggrecan Mouse Monoclonal Antibody [clone: 6-B-4] (Alexa Fluor® 647)

Supplier: Abcam
AB315140-10UL AB315140-100UL
ABCAAB315140-10ULEA 205 GBP
ABCAAB315140-10UL ABCAAB315140-100UL
Anti-Aggrecan Mouse Monoclonal Antibody [clone: 6-B-4] (Alexa Fluor® 647)
Antibodies
Alexa Fluor® 647 Mouse monoclonal [6-B-4] to Aggrecan.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Aggrecan
Clonality: Monoclonal
Clone: 6-B-4
Conjugation: Alexa Fluor® 647
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR