Anti-Squalene Epoxidase Rabbit Monoclonal Antibody [clone: EPR29195-89]

Supplier: Abcam
AB317628-20UL AB317628-100UL AB317628-1ML
ABCAAB317628-20ULEA 210 GBP
ABCAAB317628-20UL ABCAAB317628-100UL ABCAAB317628-1ML
Anti-Squalene Epoxidase Rabbit Monoclonal Antibody [clone: EPR29195-89]
Antibodies
Rabbit monoclonal [EPR29195-89] to Squalene Epoxidase.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Squalene Epoxidase
Clonality: Monoclonal
Clone: EPR29195-89
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR