Anti-EpCAM Rabbit Monoclonal Antibody [clone: EPR20532-222] (Biotin)

Supplier: Abcam
AB314939-10UL AB314939-100UL
ABCAAB314939-10ULEA 205 GBP
ABCAAB314939-10UL ABCAAB314939-100UL
Anti-EpCAM Rabbit Monoclonal Antibody [clone: EPR20532-222] (Biotin)
Antibodies
Biotin Rabbit monoclonal [EPR20532-222] to EpCAM.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: EpCAM
Clonality: Monoclonal
Clone: EPR20532-222
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR