Anti-Histone H3 Rabbit Monoclonal Antibody [clone: EPR16987]

Supplier: Abcam
AB176842-100 AB176842-1ML
ABCAAB176842-100EA 409 GBP
ABCAAB176842-100 ABCAAB176842-1ML
Anti-Histone H3 Rabbit Monoclonal Antibody [clone: EPR16987]
Antibodies
Rabbit monoclonal [EPR16987] to Histone H3 - Nuclear Marker and ChIP Grade.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Histone H3
Clonality: Monoclonal
Clone: EPR16987
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat;Saccharomyces cerevisiae;S. pombe;Drosophila melanogaster
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR