Anti-SOX9 Rabbit Monoclonal Antibody [clone: EPR14335-78]

Supplier: Abcam
AB185966-100 AB185966-1ML
ABCAAB185966-100EA 758 GBP
ABCAAB185966-100 ABCAAB185966-1ML
Anti-SOX9 Rabbit Monoclonal Antibody [clone: EPR14335-78]
Antibodies
Rabbit monoclonal [EPR14335-78] to SOX9.

Type: Primary
Antigen: SOX9
Clonality: Monoclonal
Clone: EPR14335-78
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR