Anti-HIF1 beta Rabbit Monoclonal Antibody [clone: EPR23106-151]

Supplier: Abcam
AB270520-100 AB270520-1ML
ABCAAB270520-100EA 677 GBP
ABCAAB270520-100 ABCAAB270520-1ML
Anti-HIF1 beta Rabbit Monoclonal Antibody [clone: EPR23106-151]
Antibodies
Rabbit monoclonal [EPR23106-151] to HIF1 beta.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: HIF1 beta
Clonality: Monoclonal
Clone: EPR23106-151
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR