Anti-VGF Rabbit Monoclonal Antibody [clone: EPR27029-60]

Supplier: Abcam
AB313783-100UL AB313783-20UL AB313783-1ML
ABCAAB313783-100ULEA 596 GBP
ABCAAB313783-100UL ABCAAB313783-20UL ABCAAB313783-1ML
Anti-VGF Rabbit Monoclonal Antibody [clone: EPR27029-60]
Antibodies
Rabbit monoclonal [EPR27029-60] to VGF.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: VGF
Clonality: Monoclonal
Clone: EPR27029-60
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR