Anti-BLBP Rabbit Monoclonal Antibody [clone: EPR24033-13]

Supplier: Abcam
AB279649-100 AB279649-20UL AB279649-1ML
ABCAAB279649-100EA 613 GBP
ABCAAB279649-100 ABCAAB279649-20UL ABCAAB279649-1ML
Anti-BLBP Rabbit Monoclonal Antibody [clone: EPR24033-13]
Antibodies
Rabbit monoclonal [EPR24033-13] to BLBP.

Type: Primary
Antigen: BLBP
Clonality: Monoclonal
Clone: EPR24033-13
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Human; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR