Anti-PspCas13b Mouse Monoclonal Antibody [clone: 12F2-A10]

Supplier: Abcam
AB273463-100 AB273463-1
ABCAAB273463-100EA 876 GBP
ABCAAB273463-100 ABCAAB273463-1
Anti-PspCas13b Mouse Monoclonal Antibody [clone: 12F2-A10]
Antibodies
Mouse monoclonal [12F2-A10] to PspCas13b - BSA and Azide free.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: PspCas13b
Clonality: Monoclonal
Clone: 12F2-A10
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR