Anti-Dynein light chain Rabbit Monoclonal Antibody [clone: EPR11234]

Supplier: Abcam
AB168385-100 AB168385-1ML
ABCAAB168385-100EA 575 GBP
ABCAAB168385-100 ABCAAB168385-1ML
Anti-Dynein light chain Rabbit Monoclonal Antibody [clone: EPR11234]
Antibodies
Rabbit monoclonal [EPR11234] to Dynein light chain.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Dynein light chain
Clonality: Monoclonal
Clone: EPR11234
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR