Bottle top vacuum filtration systems, Filtropur V and Filtropur BT
Supplier: SARSTEDT
83.1822 83.1822.001 83.1822.100 83.1822.101 83.1823 83.1823.001 83.1823.100 83.1823.101
SARS83.1822EA 585 GBP
SARS83.1822 SARS83.1822.001 SARS83.1822.100 SARS83.1822.101 SARS83.1823 SARS83.1823.001 SARS83.1823.100 SARS83.1823.101
Bottle top vacuum filtration systems, Filtropur V and Filtropur BT
Filters Bottle Top Filters
These units are ideal for the cold sterilisation of eg. cell culture media and protein solutions. PES 'high-flow' filter membrane for high throughput and low protein binding.
  • Non pyrogenic, non cytotoxic
  • Fast product identification by indicating the filter material, pore size and lot number on the filter housing
  • Ergonomically shaped receiver bottles
Delivery information: A sterile screw cap for the collection vessel is included with each filtration unit.
Packaging: Individually wrapped.