Chromone-3-carboxylic acid

Supplier: Molekula

MOLE11667804-1GEA 0 GBP
MOLE11667804-1G
Chromone-3-carboxylic acid
Chromone-3-carboxylic Acid
Formula: C₁₀H₆O₄
MW: 190,15 g/mol
MDL Number: MFCD00017338
CAS Number: 39079-62-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR