N-Acetylmorpholine ≥98%

Supplier: Thermo Fisher Scientific
Warning

B24120.14 B24120.22
B24120.14EA 28 GBP
B24120.14 B24120.22
N-Acetylmorpholine ≥98%
N-Acetylmorpholine

Purity: 98% CAS No.: 1696-20-4 Molecular Formula: C6H11NO2 Formula Weight: 129.16 Flash Point: 110°(230°F) Refractive Index: 1.4845 Melting Point: 9-10° Boiling Point: 245-246° Beilstein Reference: 111971 MDL No.: MFCD00006171

Formula: C₆H₁₁NO₂
Boiling Pt: 245…246 °C
Melting Pt: 9…10 °C
Density: 1,116
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006171
CAS Number: 1696-20-4
EINECS: 216-913-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR