2-Bromo-5-chloro-3-methylbenzo[b]thiophene ≥97%

Supplier: Thermo Scientific
Warning

B20250.03 B20250.06
B20250.03EA 33.9 GBP
B20250.03 B20250.06
2-Bromo-5-chloro-3-methylbenzo[b]thiophene ≥97%
2-Bromo-5-chloro-3-methylbenzo[b]thiophene

Purity: 97% CAS No.: 175203-60-8 Molecular Formula: C9H6BrClS Formula Weight: 261.57 Melting Point: 104-107° MDL No.: MFCD00126363

Formula: C₉H₆BrClS
MW: 261,57 g/mol
Melting Pt: 104…107 °C
Storage Temperature: Ambient
MDL Number: MFCD00126363
CAS Number: 175203-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR