7-Methoxyflavanone ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
B25358.06
B25358.03
B25358.06EA
0
GBP
B25358.06
B25358.03
7-Methoxyflavanone ≥98%
7-Methoxyflavanone
Purity: 98%
CAS No.: 21785-09-1
Molecular Formula: C16H14O3
Formula Weight: 254.29
Melting Point: 86-90°
MDL No.: MFCD00017486
Formula:
C₁₆H₁₄O₃ Melting Pt: 86…90 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017486 CAS Number: 21785-09-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...