4-Amino-2-(methylthio)pyrimidine-5-carbonitrile 97%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
4-Amino-5-cyano-2-(methylthio)pyrimidine
A19949.06
A19949.14
A19949.06EA
113
GBP
A19949.06
A19949.14
4-Amino-2-(methylthio)pyrimidine-5-carbonitrile 97%
4-Amino-2-(methylthio)pyrimidine-5-carbonitrile
Purity: 97%
CAS No.: 770-30-9
Molecular Formula: C6H6N4S
Formula Weight: 166.21
Melting Point: 240-243°
UN No.: 3439
MDL No.: MFCD00023237
Formula:
C₆H₆N₄S MW: 166,21 g/mol Melting Pt: 240…243 °C Storage Temperature: Ambient |
MDL Number:
MFCD00023237 CAS Number: 770-30-9 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...