2-Fluoro-L-phenylalanine ≥99%, powder

Supplier: Thermo Fisher Scientific

Synonyms: H-Phe(2-F)-OH

H27807.03 H27807.MD
H27807.03EA 0 GBP
H27807.03 H27807.MD
2-Fluoro-L-phenylalanine ≥99%, powder
2-Fluoro-L-phenylalanine

Purity: 99+% CAS No.: 19883-78-4 Molecular Formula: C9H10FNO2 Formula Weight: 183.18 MDL No.: MFCD00066448

Formula: C₉H₁₀FNO₂
Storage Temperature: Ambient
MDL Number: MFCD00066448
CAS Number: 19883-78-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR