Propyl decanoate ≥98%
Supplier: Thermo Fisher Scientific
B25159.0B
B25159.18
B25159.30
B25159.0BEA
577
GBP
B25159.0B
B25159.18
B25159.30
Propyl decanoate ≥98%
Propyl decanoate
Purity: 98%
CAS No.: 30673-60-0
Molecular Formula: CH3(CH2)8CO2CH2CH2CH3
Formula Weight: 214.35
Flash Point: 115°(239°F)
Boiling Point: 254°
MDL No.: MFCD00056329
Formula:
C₁₃H₂₆O₂ Boiling Pt: 254 °C Density: 0,86 Flash Pt: 115 °C (239 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00056329 CAS Number: 30673-60-0 EINECS: 250-283-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...