5-Benzoylpentanoic acid ≥99%

Supplier: Thermo Scientific
Warning

Synonyms: 6-Oxo-6-phenylhexanoic acid

L08290.03 L08290.06
L08290.03EA 23.4 GBP
L08290.03 L08290.06
5-Benzoylpentanoic acid ≥99%
5-Benzoylpentanoic acid

Purity: 99% CAS No.: 4144-62-1 Molecular Formula: C12H14O3 Formula Weight: 206.24 Melting Point: 75-79° Beilstein Reference: 1961325

Formula: C₁₂H₁₄O₃
MW: 206,24 g/mol
Melting Pt: 75…79 °C
Storage Temperature: Ambient
MDL Number: MFCD00014380
CAS Number: 4144-62-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR