Ferrocenemethanol 97%

Supplier: Thermo Scientific
Warning

Synonyms: Hydroxymethylferrocene

348030010 348030050 348035000
ACRO348030010EA 185 GBP
ACRO348030010 ACRO348030050 ACRO348035000
Ferrocenemethanol 97%
Ferrocenemethanol
Formula: C₁₁H₁₂FeO
MW: 216,06 g/mol
Melting Pt: 77…82 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00040878
CAS Number: 1273-86-5

Order Now

Specification Test Results

Appearance Yellow to golden to orange Powder or needles
Infrared spectrum Conforms
Melting point 77 to 82 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR