(S)-(-)-4-Amino-2-hydroxybutyric acid 96%

Supplier: Thermo Scientific
Warning

Synonyms: (S)-4-Amino-2-hydroxybutanoic acid

342430050 342430250
ACRO342430050EA 80.1 GBP
ACRO342430050 ACRO342430250
(S)-(-)-4-Amino-2-hydroxybutyric acid 96%
(S)-(-)-4-Amino-2-hydroxybutyric Acid
Formula: C₄H₉NO₃
MW: 119,12 g/mol
Melting Pt: 200…203 °C
Storage Temperature: Ambient
MDL Number: MFCD00674110
CAS Number: 40371-51-5

Order Now

Specification Test Results

Appearance White to beige Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Specific optical rotation -28° to -32° (20°C, 589 nm) (c=1, water)
Titration with HClO4 ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR