4-Nitrophenylacetone ≥98%
Supplier: Thermo Scientific
|
Warning
|
B21183.03
B21183.06
B21183.14
B21183.03EA
53.6
GBP
B21183.03
B21183.06
B21183.14
4-Nitrophenylacetone ≥98%
4-Nitrophenylacetone
Purity: 98%
CAS No.: 5332-96-7
Molecular Formula: C9H9NO3
Formula Weight: 179.18
Melting Point: 61-64°
Beilstein Reference: 1953054
Formula:
C₉H₉NO₃ MW: 179,18 g/mol Melting Pt: 61…64 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051518 CAS Number: 5332-96-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...