Potassium(bromomethyl)trifluoroborate 95%
Supplier: Thermo Fisher Scientific
|
Warning
|
H32168.03
H32168.06
H32168.03EA
0
GBP
H32168.03
H32168.06
Potassium(bromomethyl)trifluoroborate 95%
Potassium (bromomethyl)trifluoroborate
Purity: 95%
CAS No.: 888711-44-2
Molecular Formula: CH2BBrF3K
Formula Weight: 200.84
MDL No.: MFCD09265154
Formula:
CH₂BBrF₃K Storage Temperature: Ambient |
MDL Number:
MFCD09265154 CAS Number: 888711-44-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...