5-Amino-2-(trifluoromethyl)pyridine 98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
3-Amino-6-(trifluoromethyl)pyridine, 2-(Trifluoromethyl)pyridin-5-amine, 2-(Trifluoromethyl)-5-pyridylamine
H56968.MD
H56968.03
H56968.06
H56968.MDEA
41.2
GBP
H56968.MD
H56968.03
H56968.06
5-Amino-2-(trifluoromethyl)pyridine 98%
5-Amino-2-(trifluoromethyl)pyridine
Formula:
C₆H₅F₃N₂ MW: 162,11 g/mol Melting Pt: 39…43 °C Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD01862656 CAS Number: 106877-33-2 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...