Diethyl (3,4-dichlorophenyl)malonate 95%

Supplier: Thermo Scientific
Warning

H26277.03 H26277.06
H26277.03EA 59.1 GBP
H26277.03 H26277.06
Diethyl (3,4-dichlorophenyl)malonate 95%
Diethyl (3,4-dichlorophenyl)malonate
Formula: C₁₃H₁₄Cl₂O₄
MW: 305,16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02180874
CAS Number: 28751-26-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR