2-Chloro-3-cyanopyrazine

Synonyms: 3-chloropyrazine-2-carbonitrile

CFF242.2.5 CFF242.5 005877-1G 005877-5G 005877-25G
CHMPCFF242.2.5EA 0 GBP
CHMPCFF242.2.5 CHMPCFF242.5 FLUO005877-1G FLUO005877-5G FLUO005877-25G
2-Chloro-3-cyanopyrazine
2-Chloro-3-cyanopyrazine
Formula: C₅H₂ClN₃
MW: 139,5400 g/mol
MDL Number: MFCD00219653
CAS Number: 55557-52-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR