(4-Nitrophenyl) α-D-glucopyranoside

Supplier: Molekula

Synonyms: 4-Nitrophenyl α-D-glucopyranoside

MOLEM79358495EA 0 GBP
MOLEM79358495
(4-Nitrophenyl) α-D-glucopyranoside
(4-Nitrophenyl) α-D-glucopyranoside
Formula: C₁₂H₁₅NO₈
MW: 301,25 g/mol
MDL Number: MFCD00064088
CAS Number: 3767-28-0
EINECS: 223-189-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR