3-(4-Hydroxyphenyl)propiononitrile ≥98%

Supplier: Thermo Scientific
Danger

Synonyms: 3-(4-Hydroxyphenyl)propionitrile

L07934.03 L07934.06
L07934.03EA 41.2 GBP
L07934.03 L07934.06
3-(4-Hydroxyphenyl)propiononitrile ≥98%
3-(4-Hydroxyphenyl)propiononitrile

Purity: 98% CAS No.: 17362-17-3 Molecular Formula: C9H9NO Formula Weight: 147.18 Melting Point: 54-58° Boiling Point: 166°/8mm UN No.: 3439 MDL No.: MFCD00016492

Formula: C₉H₉NO
MW: 147,18 g/mol
Boiling Pt: 166 °C (8 mmHg)
Melting Pt: 54…58 °C
Storage Temperature: Ambient
MDL Number: MFCD00016492
CAS Number: 17362-17-3
EINECS: 241-393-0
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR