1-Chloro-4-(3,4-dimethylphenyl)phthalazine 96%

Supplier: Thermo Scientific
Warning

H33635.MD H33635.03 H33635.06
H33635.MDEA 20.4 GBP
H33635.MD H33635.03 H33635.06
1-Chloro-4-(3,4-dimethylphenyl)phthalazine 96%
1-Chloro-4-(3,4-dimethylphenyl)phthalazine

Purity: 96% CAS No.: 129842-38-2 Molecular Formula: C16H13ClN2 Formula Weight: 268.75 Melting Point: 151-153° MDL No.: MFCD03001385

Formula: C₁₆H₁₃ClN₂
MW: 268,75 g/mol
Melting Pt: 151…153 °C
Storage Temperature: Ambient
MDL Number: MFCD03001385
CAS Number: 129842-38-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR