4-Methoxy-1-naphthonitrile ≥98%

Supplier: Thermo Fisher Scientific

L05354.06 L05354.14
L05354.06EA 0 GBP
L05354.06 L05354.14
4-Methoxy-1-naphthonitrile ≥98%
4-Methoxy-1-naphthonitrile

Purity: 98% CAS No.: 5961-55-7 Molecular Formula: C12H9NO Formula Weight: 183.21 Melting Point: 100-103° Beilstein Reference: 2096262 UN No.: 3439 MDL No.: MFCD00003925

Formula: C₁₂H₉NO
MW: 183.2060 g/mol
MDL Number: MFCD00003925
CAS Number: 5961-55-7
EINECS: 227-734-6
UN: 3439
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR