Thiocarbohydrazide ≥97%

Supplier: Thermo Fisher Scientific
Danger

L01205.06 L01205.14 L01205.22
L01205.06EA 35.5 GBP
L01205.06 L01205.14 L01205.22
Thiocarbohydrazide ≥97%
Thiocarbohydrazide

Purity: 97% CAS No.: 2231-57-4 Molecular Formula: CH6N4S Formula Weight: 106.15 Storage Temperature: 2°C to 8°C Melting Point: ca 175° dec. Beilstein Reference: 506657 UN No.: 2811 MDL No.: MFCD00007616

Formula: CH₆N₄S
Melting Pt: ∼ 175 °C (dec.)
MDL Number: MFCD00007616
CAS Number: 2231-57-4
EINECS: 218-769-8
UN: 2811
ADR: 6.1,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR