Thiocarbohydrazide 98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: TCH

207530250 207530050 207531000
ACRO207530250EA 364 GBP
ACRO207530250 ACRO207530050 ACRO207531000
Thiocarbohydrazide 98%
Thiocarbohydrazide
Formula: CH₆N₄S
MW: 106.15 g/mol
Melting Pt: 168…172 °C
Storage Temperature: Ambient
MDL Number: MFCD00007616
CAS Number: 2231-57-4
EINECS: 218-769-8
UN: 2811
ADR: 6.1 I

Order Now

Specification Test Results

Appearance White to beige to grey Crystalline powder
Infrared spectrum Conforms
Melting point 168 to 172 °C (Dec)
Assay ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR