5-Chloro-2-methylbenzoxazole

Supplier: Molekula

Synonyms: 5-Chloro-2-methylbenzo[d]oxazole

MOLEM37075484EA 0 GBP
MOLEM37075484
5-Chloro-2-methylbenzoxazole
5-Chloro-2-methylbenzoxazole
Formula: C₈H₆ClNO
MW: 167,5900 g/mol
MDL Number: MFCD00022850
CAS Number: 19219-99-9
EINECS: 242-888-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR