2-Chloro-1-tritylimidazole ≥97%
Supplier: Thermo Scientific
|
Warning
|
H51861.MD
H51861.03
H51861.MDEA
44.7
GBP
H51861.MD
H51861.03
2-Chloro-1-tritylimidazole ≥97%
2-Chloro-1-tritylimidazole
Purity: 97%
CAS No.: 67478-48-2
Molecular Formula: C22H17ClN2
Formula Weight: 344.84
Melting Point: 200-202°
MDL No.: MFCD02179527
Formula:
C₂₂H₁₇ClN₂ MW: 344,84 g/mol Melting Pt: 200…202 °C Storage Temperature: Ambient |
MDL Number:
MFCD02179527 CAS Number: 67478-48-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...