1-Ethoxymethyl-2-iodoimidazole ≥97%

Supplier: Thermo Scientific
Warning

H51865.MD H51865.03
H51865.MDEA 51.3 GBP
H51865.MD H51865.03
1-Ethoxymethyl-2-iodoimidazole ≥97%
1-Ethoxymethyl-2-iodoimidazole

Purity: 97% CAS No.: 146697-87-2 Molecular Formula: C6H9IN2O Formula Weight: 252.05 MDL No.: MFCD03788568

Formula: C₆H₉IN₂O
MW: 252,05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03788568
CAS Number: 146697-87-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR